(±)-γ-Decalactone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Hexyldihydrofuran-2(3H)-one, Gamma-decalactone, (±)-Decan-4-olide

M-Clarity™ quality level = MQ300

W236004-1KG-K W236004-5KG-K W236004-10KG-K
SIALW236004-1KG-KEA 119 GBP
SIALW236004-1KG-K SIALW236004-5KG-K SIALW236004-10KG-K
(±)-γ-Decalactone, Sigma-Aldrich®
(±)-γ-Decalactone
Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 281 °C (1013 hPa)
Density: 0.965 g/cm³ (0 °C)
MDL Number: MFCD00005404
CAS Number: 706-14-9
EINECS: 211-892-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR