Kojic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

M-Clarity™ quality level = MQ200

K3125-10G K3125-25G K3125-5G
SIALK3125-10GEA 300 GBP
SIALK3125-10G SIALK3125-25G SIALK3125-5G
Kojic acid, Sigma-Aldrich®
Kojic acid
Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 149…152 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00006580
CAS Number: 501-30-4
EINECS: 207-922-4
Merck Index: 13,05334

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR