Iron(III) oxide ≥99.85% (metals basis)
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Ferric oxide
, Diiron trioxide
, Ferric Oxide, (iron(III) oxide)
36387.18EA
97.7
GBP
36387.18
36387.30
Iron(III) oxide ≥99.85% (metals basis)
Iron (III) oxide
Formula:
Fe₂O₃ MW: 159.69 g/mol Melting Pt: 1565 °C Density: 5.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...