Nickel(II) fluoride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Nickel difluoride
M-Clarity™ quality level = MQ100
342211-10G
342211-50G
SIAL342211-10GEA
130
GBP
SIAL342211-10G
SIAL342211-50G
Nickel(II) fluoride, Sigma-Aldrich®
Nickel (II) fluoride
Formula:
F₂Ni MW: 96.69 g/mol Melting Pt: 1360…1380 °C Density: 2.67…2.77 g/cm³ (25 °C) |
MDL Number:
MFCD00016263 CAS Number: 10028-18-9 EINECS: 233-071-3 UN: 3288 ADR: 6.1,III Merck Index: 12,06591 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...