Titanium(IV) fluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002869-25G
002869-100G
002869-250G
FLUO002869-25GEA
0
GBP
FLUO002869-25G
FLUO002869-100G
FLUO002869-250G
Titanium(IV) fluoride
Titanium (IV) fluoride
Formula:
F₄Ti MW: 123.87 g/mol Boiling Pt: 284 °C (1013 hPa) Melting Pt: 400 °C Density: 2.798 g/cm³ (20.5 °C) |
MDL Number:
MFCD00042545 CAS Number: 7783-63-3 EINECS: 232-017-6 UN: 3260 ADR: 8,II Merck Index: 12,09619 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...