Zinc bromide
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Zinc (II) bromide
ROTH9743.1EA
0
GBP
ROTH9743.1
Zinc bromide
Zinc bromide
Formula:
Br₂Zn MW: 225.2 g/mol Boiling Pt: 670…680 °C (1013 hPa) Melting Pt: 394 °C Density: 4.22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 12,10258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...