Thulium(III) oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

289167-1G 289167-5G
SIAL289167-1GEA 162 GBP
SIAL289167-1G SIAL289167-5G
Thulium(III) oxide, Sigma-Aldrich®
Thulium (III) oxide
Formula: O₃Tm₂
MW: 385.87 g/mol
Melting Pt: 2410 °C
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR