Holmium(III) oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Diholmium trioxide
, Holmium oxide
SIALH9750-50GEA
755
GBP
SIALH9750-50G
SIALH9750-10G
Holmium(III) oxide, Sigma-Aldrich®
Holmium (III) oxide
Formula:
Ho₂O₃ MW: 377.86 g/mol Boiling Pt: 2600 °C (1013 hPa) Melting Pt: 2400 °C Density: 3.79 g/cm³ (20 °C) |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 EINECS: 235-015-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...