3-Isoquinolinol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Hydroxyisoquinoline

M-Clarity™ quality level = MQ100

SIAL368954-1GEA 37.2 GBP
SIAL368954-1G
3-Isoquinolinol, Sigma-Aldrich®
3-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00075524
CAS Number: 7651-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR