4-Fluoroindole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Fluoro-1H-indole

005409-1G 005409-2.5G
FLUO005409-1GEA 0 GBP
FLUO005409-1G FLUO005409-2.5G
4-Fluoroindole
4-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD00055992
CAS Number: 387-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR