Indole-4-carbonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H-Indole-4-carbonitrile4-Cyano-1H-indole4-Cyanoindole

M-Clarity™ quality level = MQ100

SIAL645532-1GEA 39.2 GBP
SIAL645532-1G SIAL645532-5G
Indole-4-carbonitrile, Sigma-Aldrich®
Indole-4-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00152045
CAS Number: 16136-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR