Indeno[1,2,3-cd]pyrene
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
48499.EA
183
GBP
48499.
Indeno[1,2,3-cd]pyrene
Indeno[1,2,3-cd]pyrene
Formula:
C₂₂H₁₂ MW: 276.34 g/mol Boiling Pt: 536 °C (1013 hPa) Melting Pt: 163…164 °C |
CAS Number:
193-39-5 EINECS: 205-893-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...