Indeno[1,2,3-cd]pyrene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

EHERC20830000EA 425 GBP
EHERC20830000
Indeno[1,2,3-cd]pyrene
Indeno[1,2,3-cd]pyrene
Formula: C₂₂H₁₂
MW: 276.34 g/mol
Boiling Pt: 536 °C (1013 hPa)
Melting Pt: 163…164 °C
CAS Number: 193-39-5
EINECS: 205-893-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR