5-Fluoro-1-indanone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Fluoro-2,3-dihydro-1H-inden-1-one

001181-5G 001181-100G 001181-25G
FLUO001181-5GEA 0 GBP
FLUO001181-5G FLUO001181-100G FLUO001181-25G
5-Fluoro-1-indanone
5-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
Boiling Pt: 113…114 °C (10 mmHg)
Melting Pt: 35…37 °C
Density: 1,35 g/cm³ (20 °C)
MDL Number: MFCD00041031
CAS Number: 700-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR