Diphenyl sulphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
P35316-100G
P35316-25G
SIALP35316-100GEA
169
GBP
SIALP35316-100G
SIALP35316-25G
Diphenyl sulphide, Sigma-Aldrich®
Diphenyl sulphide
Formula:
C₁₂H₁₀S Boiling Pt: 296 °C (1013 hPa) Melting Pt: –40 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003064 CAS Number: 139-66-2 EINECS: 205-371-4 UN: 2810 ADR: 6.1,III Merck Index: 12,07467 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...