Diphenyl disulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

169021-250G 169021-50G 169021-10G
SIAL169021-250GEA 219 GBP
SIAL169021-250G SIAL169021-50G SIAL169021-10G
Diphenyl disulphide, Sigma-Aldrich®
Diphenyl disulphide
Formula: C₁₂H₁₀S₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 58…60 °C
Density: 1.155 g/cm³ (64.9 °C)
MDL Number: MFCD00003065
CAS Number: 882-33-7
EINECS: 212-926-4
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR