Diphenyl disulphide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002923-100G
002923-25G
FLUO002923-100GEA
0
GBP
FLUO002923-100G
FLUO002923-25G
Diphenyl disulphide
Diphenyl disulphide
Formula:
C₁₂H₁₀S₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 58…60 °C Density: 1.155 g/cm³ (64.9 °C) |
MDL Number:
MFCD00003065 CAS Number: 882-33-7 EINECS: 212-926-4 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...