Nitrosium hexafluoroantimonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Nitrosyl hexafluoroantimonate, Nitrosonium hexafluoroantimonate

006897-10G 006897-50G
FLUO006897-10GEA 0 GBP
FLUO006897-10G FLUO006897-50G
Nitrosium hexafluoroantimonate
Nitrosium hexafluoroantimonate
Formula: F₆NOSb
MW: 265.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042550
CAS Number: 16941-06-3
UN: 1549
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR