Butyrylthiocholine chloride

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLEM70336609EA 0 GBP
MOLEM70336609
Butyrylthiocholine chloride
Butyrylthiocholine chloride
Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃Cl
MW: 225.78 g/mol
Storage Temperature: Refrigerator
CAS Number: 22026-63-7
EINECS: 244-731-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR