Tetrabutylammonium tribromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

M-Clarity™ quality level = MQ200

301590-100G 301590-25G
SIAL301590-100GEA 97.5 GBP
SIAL301590-100G SIAL301590-25G
Tetrabutylammonium tribromide, Sigma-Aldrich®
Tetrabutylammonium tribromide
Formula: C₁₆H₃₆Br₃N
MW: 482.18 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00012110
CAS Number: 38932-80-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR