Tetraethylammonium bromide

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tetrylammonium bromide

ARC-577 ARC-577-1
BTRNARC-577EA 0 GBP
BTRNARC-577 BTRNARC-577-1
Tetraethylammonium bromide
Tetraethylammonium bromide
Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: > 285 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR