Phenyltrimethoxysilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Trimethoxy(phenyl)silane
M-Clarity™ quality level = MQ200
8.43658.0050EA
81.6
GBP
8.43658.0050
Phenyltrimethoxysilane for synthesis, Sigma-Aldrich®
Phenyltrimethoxysilane
Formula:
C₉H₁₄O₃Si MW: 198.29 g/mol Density: 1.064 g/cm³ (20 °C) |
MDL Number:
MFCD00025689 CAS Number: 2996-92-1 EINECS: 221-066-9 |
Specification Test Results
Molar Mass | 198.3 g/mol |
Boiling Point | 233 °C |
Flash Point | 83 °C |
Density | 1.064 g/cm3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...