Hexabutylditin, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Hexa-n-butylditin, Hexabutyldistannane, Bis(tributyltin)
M-Clarity™ quality level = MQ100
251127-10G
251127-50G
SIAL251127-10GEA
102
GBP
SIAL251127-10G
SIAL251127-50G
Hexabutylditin, Sigma-Aldrich®
Hexabutylditin
Formula:
C₂₄H₅₄Sn₂ MW: 580.11 g/mol Boiling Pt: 160 °C (0.8 hPa) Density: 1.162 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009417 CAS Number: 813-19-4 EINECS: 212-383-3 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...