Tetrabutyltin ≥93%, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Tetrabutylstannane, Tetra-n-butyltin
M-Clarity™ quality level = MQ100
T6008-100G-A
T6008-5G-A
SIALT6008-100G-AEA
87
GBP
SIALT6008-100G-A
SIALT6008-5G-A
Tetrabutyltin ≥93%, Technical Grade, Sigma-Aldrich®
Tetrabutyltin
Formula:
C₁₆H₃₆Sn MW: 347.17 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –97 °C Density: 1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009422 CAS Number: 1461-25-2 EINECS: 215-960-8 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...