Trioctylsilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

448923-25ML 448923-5ML
SIAL448923-25MLEA 380 GBP
SIAL448923-25ML SIAL448923-5ML
Trioctylsilane, Sigma-Aldrich®
Trioctylsilane
Formula: C₂₄H₅₂Si
MW: 368.76 g/mol
MDL Number: MFCD00053812
CAS Number: 18765-09-8
EINECS: 242-559-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR