Dibutyltin oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide
M-Clarity™ quality level = MQ100
34980-100G
34980-500G
SIAL34980-100GEA
67.5
GBP
SIAL34980-100G
SIAL34980-500G
Dibutyltin oxide, Sigma-Aldrich®
Dibutyltin oxide
Formula:
C₈H₁₈OSn MW: 248.94 g/mol Melting Pt: > 300 °C Density: 1.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001992 CAS Number: 818-08-6 EINECS: 212-449-1 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...