Dibutyltin oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

M-Clarity™ quality level = MQ100

183083-100G 183083-500G
SIAL183083-100GEA 78.6 GBP
SIAL183083-100G SIAL183083-500G
Dibutyltin oxide, Sigma-Aldrich®
Dibutyltin oxide
Formula: C₈H₁₈OSn
MW: 248.94 g/mol
Melting Pt: > 300 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR