(3-Mercaptopropyl)triethoxysilane

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Triethoxysilyl)propanethiol3-Mercaptopropyltriethoxysilane

ABCRAB111218.0025EA 0 GBP
ABCRAB111218.0025
(3-Mercaptopropyl)triethoxysilane
(3-Mercaptopropyl)triethoxysilane
Formula: HSH₂CCH₂CH₂Si(OCH₂CH₃)₃
MW: 238.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00053924
CAS Number: 14814-09-6
EINECS: 238-883-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR