Sodium hexafluorophosphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

208051-10G 208051-250G 208051-50G
SIAL208051-10GEA 103 GBP
SIAL208051-10G SIAL208051-250G SIAL208051-50G
Sodium hexafluorophosphate, Sigma-Aldrich®
Sodium hexafluorophosphate
Formula: F₆NaP
MW: 167.95 g/mol
Melting Pt: 450 °C
Density: 2.505 g/cm³ (20 °C)
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR