Lithium fluoride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

SIAL669431-50GEA 262 GBP
SIAL669431-50G
Lithium fluoride, Sigma-Aldrich®
Lithium fluoride
Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 870 °C
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR