Potassium tetrahydroborate
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Potassium tetrahydridoborate
, Potassium borohydride
MOLE53642148-100GEA
0
GBP
MOLE53642148-100G
Potassium tetrahydroborate
Potassium tetrahydroborate
Formula:
H₄BK MW: 53.94 g/mol Melting Pt: 500 °C Density: 1.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011396 CAS Number: 13762-51-1 EINECS: 237-360-5 UN: 1870 ADR: 4.3,I Merck Index: 12,07777 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...