Sodium tetraethylborate ≥98%
Supplier: Strem Chemicals, Inc.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
STRE11-0575-5GEA
0
GBP
STRE11-0575-5G
ABCRS11-0575.0001
ABCRS11-0575
Sodium tetraethylborate ≥98%
Sodium tetraethylborate
Formula:
C₈H₂₀BNa MW: 150.05 g/mol Melting Pt: Dec. |
MDL Number:
MFCD00061547 CAS Number: 15523-24-7 UN: 3393 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...