Propylene carbonate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

D4645S D4645R D4645P D4645M D4645L
ROMID4645SEA 114 GBP
ROMID4645S ROMID4645R ROMID4645P ROMID4645M ROMID4645L
Propylene carbonate
Propylene carbonate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 241…242 °C (1013 hPa)
Melting Pt: –48.8 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 132 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR