2,4,6,8-Tetramethylcyclotetrasiloxane ≥99%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,3,5,7-Tetramethylcyclotetrasiloxane
L16642.06
L16642.14
L16642.22
L16642.06EA
27.3
GBP
L16642.06
L16642.14
L16642.22
2,4,6,8-Tetramethylcyclotetrasiloxane ≥99%
2,4,6,8-Tetramethylcyclotetrasiloxane
Purity: 99%
CAS No.: 2370-88-9
Molecular Formula: C4H16O4Si4
Formula Weight: 240.51
Flash Point: 25°(77°F)
Refractive Index: 1.3870
Melting Point: -69°
Boiling Point: 134-135°
Beilstein Reference: 1787074
UN No.: 1993
MDL No.: MFCD00039567
Formula:
C₄H₁₆O₄Si₄ MW: 240.51 g/mol Boiling Pt: 134…135 °C Melting Pt: –69 °C Density: 0.991 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039567 CAS Number: 2370-88-9 EINECS: 219-137-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...