3-Amino-2-cyclohexen-1-one
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E2181.10
E2181.100
E2181.25
MENAE2181.10EA
0
GBP
MENAE2181.10
MENAE2181.100
MENAE2181.25
3-Amino-2-cyclohexen-1-one
3-Amino-2-cyclohexen-1-one
Formula:
C₆H₉NO MW: 111.14 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: 60 °C |
MDL Number:
MFCD00013783 CAS Number: 5220-49-5 EINECS: 226-014-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...