Cyclobutylméthanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cyclobutanemethanol, Cyclobutylmethanol, Hydroxymethylcyclobutane

009111-25G 009111-100G 009111-5G
FLUO009111-25GEA 0 GBP
FLUO009111-25G FLUO009111-100G FLUO009111-5G
Cyclobutylméthanol
Cyclobutylméthanol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 143 °C (1013 hPa)
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001330
CAS Number: 4415-82-1
EINECS: 224-575-4
UN: 1105
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR