Nonafluoro(trifluoromethyl)cyclopentane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007090-100G 007090-500G 007090-25G
FLUO007090-100GEA 0 GBP
FLUO007090-100G FLUO007090-500G FLUO007090-25G
Nonafluoro(trifluoromethyl)cyclopentane
Nonafluoro(trifluoromethyl)cyclopentane
Formula: C₆F₁₂
MW: 300.05 g/mol
MDL Number: MFCD00066611
CAS Number: 1805-22-7
EINECS: 217-298-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR