myo-Inositol

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Inositolrel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaoli-InositolD-Myo-Inositol

MOLEM85563331EA 0 GBP
MOLEM85563331
myo-Inositol
myo-Inositol
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR