β-BHC

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: β-HCH(1a,2b,3a,4b,5a,6b)-1,2,3,4,5,6-Hexachlorocyclohexane

48494.EA 67.6 GBP
48494.
β-BHC
β-BHC
Formula: C₆H₆Cl₆
MW: 290.83 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 297 °C
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 319-85-7
EINECS: 206-271-3
UN: 2761
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR