β-BHC
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
β-HCH
, (1a,2b,3a,4b,5a,6b)-1,2,3,4,5,6-Hexachlorocyclohexane
EHERC14072000EA
182
GBP
EHERC14072000
β-BHC
β-BHC
Formula:
C₆H₆Cl₆ MW: 290.83 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 297 °C Density: 1.89 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
319-85-7 EINECS: 206-271-3 UN: 2761 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...