Bromocyclobutane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cyclobutyl bromide
M-Clarity™ quality level = MQ100
226998-1G
226998-5G
SIAL226998-1GEA
92.6
GBP
SIAL226998-1G
SIAL226998-5G
Bromocyclobutane, Sigma-Aldrich®
Bromocyclobutane
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 109…113 °C (1013 hPa) Density: 1.4208 g/cm³ (20 °C) |
MDL Number:
MFCD00001317 CAS Number: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...