Vitamin B12 (cyanocobalamin)
Supplier: Oxford Biotechnology
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CHMCCBL64EA
0
GBP
CHMCCBL64
OXBTCBL64
Vitamin B12 (cyanocobalamin)
Vitamin B12 (cyanocobalamin)
Formula:
C₆₃H₈₈CoN₁₄O₁₄P MW: 1355.38 g/mol Melting Pt: 320 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151092 CAS Number: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...