Benzo[ghi]perylene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
B9009-100MG-A
B9009-25MG-A
SIALB9009-100MG-AEA
298
GBP
SIALB9009-100MG-A
SIALB9009-25MG-A
Benzo[ghi]perylene, Sigma-Aldrich®
Benzo[ghi]perylene
Formula:
C₂₂H₁₂ MW: 276.34 g/mol Boiling Pt: 500 °C (1013 hPa) Melting Pt: 278…280 °C |
MDL Number:
MFCD00004135 CAS Number: 191-24-2 EINECS: 205-883-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...