Biphenyl-4-ol ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: [1,1'-Biphenyl]-4-ol, 4-Hydroxydiphenyl, 4-Phenylphenol, p-Hydroxybiphenyl, 4-Hydroxybiphenyl

A10817.0E A10817.22 A10817.36
A10817.0EEA 380 GBP
A10817.0E A10817.22 A10817.36
Biphenyl-4-ol ≥98%
Biphenyl-4-ol

Purity: 98% CAS No.: 92-69-3 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 165°(329°F) Melting Point: 164-168° Boiling Point: 305-308° Beilstein Reference: 1907452 Merck Reference: 14,7305 UN No.: 3077 MDL No.: MFCD00002347

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 305…308 °C
Melting Pt: 164…168 °C
Density: 1.29 g/cm³ (20 °C)
Flash Pt: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002347
CAS Number: 92-69-3
EINECS: 202-179-2
ADR: 9,III
Merck Index: 14,7305

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR