Biphenyl-2-ol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Phenylphenol, 2-Hydroxydiphenyl, 2-Hydroxybiphenyl

M-Clarity™ quality level = MQ200

P28263-500G P28263-2KG
SIALP28263-500GEA 97.2 GBP
SIALP28263-500G SIALP28263-2KG
Biphenyl-2-ol, Sigma-Aldrich®
Biphenyl-2-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 280…286 °C (1013 hPa)
Melting Pt: 56…58 °C
Density: 1.213 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00002208
CAS Number: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9,III
Merck Index: 12,07458

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR