2-Fluorobiphenyl

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1

001705-25G 001705-100G
FLUO001705-25GEA 0 GBP
FLUO001705-25G FLUO001705-100G
2-Fluorobiphenyl
2-Fluorobiphenyl
Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 73…74 °C
Density: 1.2452 g/cm³ (25 °C)
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR