3,3'-Dichlorobiphenyl (PCB No. 11)
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC20001100EA
85
GBP
EHERC20001100
3,3'-Dichlorobiphenyl (PCB No. 11)
3,3'-Dichlorobiphenyl (PCB No. 11)
Formula:
C₁₂H₈Cl₂ MW: 223.1 g/mol |
CAS Number:
2050-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...