3,3'-Dichlorobiphenyl (PCB No. 11)

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PROOCIL-PCB-11-CEA 1010 GBP
PROOCIL-PCB-11-C
3,3'-Dichlorobiphenyl (PCB No. 11)
3,3'-Dichlorobiphenyl (PCB No. 11)
Formula: C₁₂H₈Cl₂
MW: 223.1 g/mol
CAS Number: 2050-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR