2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)

Supplier: ACCUSTANDARD
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

122482YEA 0 GBP
122482Y
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
Formula: C₁₂H₅Cl₅
MW: 326.44 g/mol
CAS Number: 31508-00-6
UN: 2315
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR