2,4,6-Triisopropylbenzenesulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2,4,6-Triisopropylbenzene-1-sulphonyl chloride
003615-25G
003615-100G
FLUO003615-25GEA
0
GBP
FLUO003615-25G
FLUO003615-100G
2,4,6-Triisopropylbenzenesulphonyl chloride
2,4,6-Triisopropylbenzenesulphonyl chloride
Formula:
C₁₅H₂₃ClO₂S MW: 302.87 g/mol Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007433 CAS Number: 6553-96-4 EINECS: 229-479-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...